Learn More
Sino Biological Anti-Dectin-1/CLEC7A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50233RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CLEC7A extracellular domain (rM CLEC7A; Catalog#50233-M07H; Phe 69-Leu 244; NP_064392.2). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3354