Learn More
Bon Opus Biosciences Recombinant Mouse Clec9A (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP106550UG
Encompass_Preferred
Recombinant Mouse Clec9A (C-mFc) is produced by Human Cells. The target gene encoding Met1-ILe238 is expressed with a C-mFc tag. Accession# Q8BRU4. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1065
Catalog No. 50-217-2663