Learn More
Sino Biological Recombinant Anti-Clusterin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50485R10220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Clusterin/Apolipoprotein J/Apo-J (rM Clusterin/Apolipoprotein J/Apo-J; Catalog#50485-M08H; NP_038520.2; Met1-Glu448).
Catalog No. 50-240-8164