Learn More
Sino Biological Anti-Clusterin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50485RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Clusterin / Apolipoprotein J / Apo-J / CLU (rM Clusterin / Apolipoprotein J / Apo-J / CLU; Catalog#50485-M08H; NP_038520.2; Met 1-Glu 448). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1818