Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 146 (CD146) CLIA Kit
Supplier:  Biomatik Corporation EKU09777
Encompass_Preferred
Mouse Cluster Of Differentiation 146 (CD146) CLIA Kit for detecting CD146 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 54.9-40,000pg/ml. Sensitivity: 22.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q8R2Y2. Gene ID: 84004. Target Synonyms: MCAM; MUC18; Melanoma Cell Adhesion Molecule; Cell surface glycoprotein P1H12; Melanoma-associated antigen A32; S-endo 1 endothelial-associated antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6257