Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 64 (CD64) ELISA Kit
Supplier:  Biomatik Corporation EKU07026
Encompass_Preferred
Mouse Cluster Of Differentiation 64 (CD64) ELISA Kit for detecting CD64 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.257ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26151. Gene ID: 14129. Target Synonyms: FcgRI; FCGR1; FCGR1A; FCGR1B; FCGR1C; FcgRIA; CD64A; FcgRIB; CD64B; FcgRIC; CD64C; Receptor I For The Fc Region Of Immunoglobulin G; High affinity immunoglobulin gamma Fc receptor I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7560