Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 19 (CD19) ELISA Kit
Supplier:  Biomatik Corporation EKU03229
Encompass_Preferred
Mouse Cluster Of Differentiation 19 (CD19) ELISA Kit for detecting CD19 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25918. Gene ID: 12478. Target Synonyms: B4; B-lymphocyte surface antigen B4; T-cell surface antigen Leu-12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7565