Learn More
Biomatik Corporation Mouse Cluster Of differentiation 301 (CD301) ELISA Kit
Supplier:  Biomatik Corporation EKU10761
Encompass_Preferred
Mouse Cluster Of differentiation 301 (CD301) ELISA Kit for detecting CD301 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49300. Gene ID: 17312. Target Synonyms: CLEC10A; CLEC10-A; CLECSF13; CLECSF14; C-Type Lectin Domain Family 10, Member A; Macrophage Lectin 2 (Calcium Dependent) ; C-type lectin superfamily member 14; Macrophage lectin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8294