Learn More
Biomatik Corporation Mouse Cluster Of Differentiation (CD163) ELISA Kit
Supplier:  Biomatik Corporation EKU03220
Encompass_Preferred
Mouse Cluster Of Differentiation (CD163) ELISA Kit for detecting CD163 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2VLH6. Gene ID: 93671. Target Synonyms: M130; MM130; Scavenger receptor cysteine-rich type 1 protein M130; Hemoglobin scavenger receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9056