Learn More
Biomatik Corporation Mouse Cluster of Differentiation 90 (CD90) ELISA Kit
Supplier:  Biomatik Corporation EKU07653
Encompass_Preferred
Mouse Cluster of Differentiation 90 (CD90) ELISA Kit for detecting CD90 in serum, plasma, urine, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.50-800pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01831. Gene ID: 21838. Target Synonyms: Thy1; CDw90; THY mocyte differentiation antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8739