Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 23 (CD23) ELISA Kit
Supplier:  Biomatik Corporation EKU07028
Encompass_Preferred
Mouse Cluster Of Differentiation 23 (CD23) ELISA Kit for detecting CD23 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.66pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20693. Gene ID: 14128. Target Synonyms: FceRII; FCER2; BLAST-2; CLEC4J; FCE2; IGEBF; Receptor II For The Fc Region Of Immunoglobulin E; C-type lectin domain family 4 member J; Immunoglobulin E-binding factor; Lymphocyte IgE receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8784