Learn More
Biomatik Corporation Mouse Cluster of Differentiation 59 (CD59) ELISA Kit
Supplier:  Biomatik Corporation EKU06890
Encompass_Preferred
Mouse Cluster of Differentiation 59 (CD59) ELISA Kit for detecting CD59 in serum, plasma, urine, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.50-4000pg/ml. Sensitivity: 25.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O55186. Gene ID: 12509. Target Synonyms: Protectin; MIC11; MIN1; MIN2; MIN3; MSK21; MAC-IP; MACIF; MIRL; HRF20; Complement Regulatory Protein; 20 kDa homologous restriction factor; Membrane attack complex inhibition factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5777