Learn More
Raybiotech Inc Mouse Clusterin ELISA Kit
Supplier:  Raybiotech Inc ELMCLUSTERIN1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse Clusterin.
Catalog No. 50-194-9950