Learn More
Sino Biological Recombinant Mouse c-MET/HGFR Protein (hFc Tag), HPLC-verified 1
Supplier:  Sino Biological 50622M02H1
Encompass_Preferred
A DNA sequence encoding the mouse MET (NP_032617.2) extracellular domain (Met 1-Asn 929) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7186