Learn More
Sino Biological Recombinant Anti-CNDP2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50246R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CNDP2 (rM CNDP2; Catalog#50246-M08B; NP_075638.2; Met 1-Asn 475).
Catalog No. 50-255-3345