Learn More
Sino Biological Anti-CNDP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50246RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CNDP2 (rM CNDP2; Catalog#50246-M08B; NP_075638.2; Met 1-Asn 475). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3343