Learn More
Sino Biological Anti-CNTF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51181T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CNTF / Ciliary Neurotrophic Factor (rM CNTF / Ciliary Neurotrophic Factor; Catalog#51181-M07E; NP_740756.1; Ala2-Met198). CNTF / Ciliary Neurotrophic Factor specific IgG was purified by Mouse CNTF / Ciliary Neurotrophic Factor affinity chromatography.
Catalog No. 50-241-2126