Learn More
Sino Biological Anti-Contactin-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50933T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Contactin 1 / CNTN1 (rM Contactin 1 / CNTN1; Catalog#50933-M08H; NP_031753.1; Met1-Leu1000). Contactin 1 / CNTN1 specific IgG was purified by Mouse Contactin 1 / CNTN1 affinity chromatography.
Catalog No. 50-241-1934