Learn More
Sino Biological Anti-TAG1/Contactin 2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51111T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Contactin 2 / CNTN2 (Catalog#51111-M08H; Q61330; Gln31-Glu1013). Contactin 2 / CNTN2 specific IgG was purified by Mouse Contactin 2 / CNTN2 affinity chromatography.
Catalog No. 50-240-9607