Learn More
Sino Biological Anti-CASPR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50649RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CNTNAP2 / CASPR2 (rM CNTNAP2 / CASPR2; Catalog#50649-M08H; NP_001004357.2; Met 1-Ser 1262). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3168