Learn More
Sino Biological Anti-CASPR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50649T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CASPR2 / CNTNAP2 (rM CASPR2 / CNTNAP2; Catalog#50649-M08H; NP_001004357.2; Met1-Ser1262). CASPR2 / CNTNAP2 specific IgG was purified by Mouse CASPR2 / CNTNAP2 affinity chromatography.
Catalog No. 50-255-3166