Learn More
ACROBiosystems Mouse Coagulation Factor III / Tissue Factor Protein, His Tag
Catalog No. 502054714
Encompass_Preferred
Recombinant Protein;100UG;Mouse Tissue Factor, His Tag (TF3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Glu 251 (Accession # P20352-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.3 kDa.
Catalog No. 50-205-4714 Supplier ACROBiosystems Supplier No. TF3M52H3100UG
Recombinant Protein;100UG;Mouse Tissue Factor, His Tag (TF3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Glu 251 (Accession # P20352-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.3 kDa.