Learn More
American Research Products Inc Mouse coagulation factor V FV
Catalog No. 5000327747
Encompass
Mouse coagulation factor V FV ELISA Kit from CUSABIO Detection Range 78 ng/mL-5000 ng/mL Sensitivity 19 5 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27747 Supplier American Research Products Inc Supplier No. CSBE17815M
Mouse coagulation factor V FV ELISA Kit from CUSABIO Detection Range 78 ng/mL-5000 ng/mL Sensitivity 19 5 ng/mL Sample serum plasma Method Sandwich