Learn More
American Research Products Inc Mouse coagulation factor VIII
Catalog No. 5000329295
Encompass
Mouse coagulation factor (F ) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29295 Supplier American Research Products Inc Supplier No. CSBE13084M
Mouse coagulation factor (F ) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich