Learn More
Biomatik Corporation Mouse Coagulation Factor X (F10) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11627
Encompass_Preferred
Mouse Coagulation Factor X (F10) Mini sample ELISA Kit for detecting F10 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88947. Gene ID: 14058. Target Synonyms: FX; FXA; Stuart Prower Factor; Thrombokinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8117