Learn More
American Research Products Inc Mouse coagulation factor X FX
Catalog No. 5000329604
Encompass
Mouse coagulation factor X FX ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29604 Supplier American Research Products Inc Supplier No. CSBE08442M
Mouse coagulation factor X FX ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma Method Sandwich