Learn More
American Research Products Inc Mouse Cold inducible RNA bindi
Catalog No. 5000327820
Encompass
Mouse Cold-inducible RNA-binding protein (CIRBP) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27820 Supplier American Research Products Inc Supplier No. CSBEL005440MO
Mouse Cold-inducible RNA-binding protein (CIRBP) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich