Learn More
American Research Products Inc Mouse Collagen alpha 1 I cha
Catalog No. 5000327927
Encompass
Mouse Collagen alpha-1 (I) chain (COL1A1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27927 Supplier American Research Products Inc Supplier No. CSBEL005727MO
Mouse Collagen alpha-1 (I) chain (COL1A1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich