Learn More
Biomatik Corporation Mouse Colony Stimulating Factor 3, Granulocyte (GCSF) CLIA Kit
Supplier:  Biomatik Corporation EKU09751
Encompass_Preferred
Mouse Colony Stimulating Factor 3, Granulocyte (GCSF) CLIA Kit for detecting GCSF in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.58pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09920. Gene ID: 12985. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6285