Learn More
Biomatik Corporation Mouse Colony Stimulating Factor 1, Macrophage (MCSF) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11088
Encompass_Preferred
Mouse Colony Stimulating Factor 1, Macrophage (MCSF) Mini sample ELISA Kit for detecting MCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07141. Gene ID: 12977. Target Synonyms: CSF1; M-CSF; Lanimostim; Processed macrophage colony-stimulating factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6512