Learn More
American Research Products Inc Mouse Complement C1q subcompon
Catalog No. 5000327031
Encompass
Mouse Complement C1q subcomponent subunit A (C1QA) ELISA kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 039 g/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27031 Supplier American Research Products Inc Supplier No. CSBEL003637MO
Mouse Complement C1q subcomponent subunit A (C1QA) ELISA kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 039 g/mL Sample serum plasma tissue homogenates Method Sandwich