Learn More
American Research Products Inc Mouse Complement C1q tumor nec
Catalog No. 5000327764
Encompass
Mouse Complement C1q tumor necrosis factor-related protein 3 (C1QTNF3) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27764 Supplier American Research Products Inc Supplier No. CSBEL003648MO
Mouse Complement C1q tumor necrosis factor-related protein 3 (C1QTNF3) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich