Learn More
Abcam Mouse Complement C5a ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1937181X96TESTS
Encompass_Preferred
Mouse Complement C5a ELISA Kit ab193718 is used to quantify Mouse Complement C5a with a sensitivity of 6 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2168370