Learn More
Biomatik Corporation Mouse Complement Component 3a (C3a) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12352
Encompass_Preferred
Mouse Complement Component 3a (C3a) High-sensitive ELISA Kit for detecting C3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01027. Gene ID: 12266. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7670