Learn More
Biomatik Corporation Mouse Complement Component 4 (C4) CLIA Kit
Supplier:  Biomatik Corporation EKU12537
Encompass_Preferred
Mouse Complement Component 4 (C4) CLIA Kit for detecting C4 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01029. Gene ID: 12268. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6204