Learn More
Biomatik Corporation Mouse Complement Component 5a (C5a) CLIA Kit
Supplier:  Biomatik Corporation EKU08867
Encompass_Preferred
Mouse Complement Component 5a (C5a) CLIA Kit for detecting C5a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P06684. Gene ID: 15139. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7522