Learn More
American Research Products Inc Mouse complement factor H CFH
Catalog No. 5000329527
Encompass
Mouse complement factor H CFH ELISA Kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29527 Supplier American Research Products Inc Supplier No. CSBE08933M
Mouse complement factor H CFH ELISA Kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma tissue homogenates Method Sandwich