Learn More
American Research Products Inc Mouse Complement factor I CFI
Catalog No. 5000327731
Encompass
Mouse Complement factor I (CFI) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27731 Supplier American Research Products Inc Supplier No. CSBEL005279MO
Mouse Complement factor I (CFI) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich