Learn More
American Research Products Inc Mouse Complement fragment 3a
Catalog No. 5000329111
Encompass
Mouse Complement fragment 3a C3a ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29111 Supplier American Research Products Inc Supplier No. CSBE08511M
Mouse Complement fragment 3a C3a ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich