Learn More
American Research Products Inc Mouse Complement fragment 4a
Catalog No. 5000329624
Encompass
Mouse Complement fragment 4a C4a ELISA Kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29624 Supplier American Research Products Inc Supplier No. CSBE08517M
Mouse Complement fragment 4a C4a ELISA Kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates Method Sandwich