Learn More
Cayman Chemical Mouse ConstItutIve ndrostn 96
SDP
Catalog No. 502967772
Encompass_Preferred
mCAR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Item Nos. M00901-32, M00901]
Catalog No. 50-296-7772 Supplier Cayman Chemical Supplier No. 1881296 WELL
mCAR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Item Nos. M00901-32, M00901]