Learn More
Biomatik Corporation Mouse Contactin 1 (CNTN1) CLIA Kit
Supplier:  Biomatik Corporation EKU09516
Encompass_Preferred
Mouse Contactin 1 (CNTN1) CLIA Kit for detecting CNTN1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P12960. Gene ID: 12805. Target Synonyms: F3; Glycoprotein gP135; Neural cell surface protein F3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8506