Learn More
Crystal Chem Inc Mouse Corticosterone ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80556
Encompass
The Mouse Corticosterone ELISA is used to quantify levels of mouse corticosterone in serum and plasma using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 15 - 2250 ng/mL. No extraction needed, ready to use reagents. High sensitivity of 6.1 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7728