Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative determination of IgG Antibodies to SARS-CoV-2 Spike Protein in mouse serum and plasma. Assay Range: 15 - 720 ng/ml; Sensitivity: 12 ng/ml.
Catalog No. NC2747089
Encompass_Preferred
Pack Size: 1 x 96 wellsBackground:The GENLISA ELISA kits are used for assessing the specific biomarker in samples analytes which may be mouse serum, plasma and cell culture supernatant as validated with the kit. This ELISA measures IgG antibodies to SARS-CoV-2 Spike Protein RBD antigen specifically. This SARS-CoV-2 ELISA kit employs a sandwich ELISA technique which leads to a higher specificity and increased sensitivity compared to conventional competitive ELISA kits which employ only one antibody. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.
Catalog No. NC2747089 Supplier KRISHGEN BIOSYSTEMS Supplier No. KBVH01513
Pack Size: 1 x 96 wellsBackground:The GENLISA ELISA kits are used for assessing the specific biomarker in samples analytes which may be mouse serum, plasma and cell culture supernatant as validated with the kit. This ELISA measures IgG antibodies to SARS-CoV-2 Spike Protein RBD antigen specifically. This SARS-CoV-2 ELISA kit employs a sandwich ELISA technique which leads to a higher specificity and increased sensitivity compared to conventional competitive ELISA kits which employ only one antibody. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.