Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative determination of IgG Antibodies to SARS-CoV-2 Spike S1+S2 protein in mouse serum and plasma. Assay Range: 15 - 720 ng/ml; Sensitivity: 12 ng/ml
Supplier:  KRISHGEN BIOSYSTEMS KBVH01515
Encompass_Preferred
Pack Size: 1 x 96 wells. Introduction: The GENLISA ELISA kits are used for assessing the specific biomarker in samples analytes which may be serum, plasma and cell culture supernatant as validated with the kit. The kit employs a sandwich ELISA technique which leads to a higher specificity and increased sensitivity compared to conventional competitive ELISA kits which employ only one antibody. Principle: The method employs indirect sandwich ELISA technique. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <10%.
Catalog No. NC2747081