Learn More
Sino Biological Anti-Carboxypeptidase A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50448RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CPA1 (rM CPA1; Catalog#50448-M08H; NP_079626.2; Met 1-Tyr 419). CPA1 specific IgG was purified by mouse CPA1 affinity chromatography.
Catalog No. 50-241-1786