Learn More
Sino Biological Anti-Carboxypeptidase A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50448RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CPA1 (rM CPA1; Catalog#50448-M08H; NP_079626.2; Met 1-Tyr 419). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1785