Learn More
Sino Biological Mouse CPA1 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50448M08H50
Encompass_Preferred
A DNA sequence encoding the mouse CPA1 (NP_079626.2) (Met 1-Tyr 419) precusor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-237-3218