Learn More
Sino Biological Anti-Carboxypeptidase A2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50778RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Carboxypeptidase A2 / CPA2 (Catalog#50778-M08H; Q504N0; Met1-Tyr417). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7718