Learn More
Sino Biological Mouse CPB1 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50386M08H50
Encompass_Preferred
A DNA sequence encoding the mouse CPB1 (NP_083982.1) precursor (Met 1-Tyr 415) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-237-0505